1,2,3,3-Tetramethyl-3H-indolium iodide ≥98%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

328979-25G
328979-25GEA 259 EUR
328979-25G
1,2,3,3-Tetramethyl-3H-indolium iodide ≥98%, Sigma-Aldrich®
1,2,3,3-Tetramethyl-3H-indolium iodide

1,2,3,3-Tetramethyl-3H-indolium iodide,98%

Formel: C₁₂H₁₆IN
Molecular Weight: 301,17 g/mol
Schmelzpunkt: 258 °C (Zers., Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00013333
CAS-Nummer: 5418-63-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ee7f05cba8cda21ccb6fad0aa8d7d2f85a2fcff&vid=bfcd4a54ae830b908538139d8f03a5c0238feb3d&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR