Organic reference standard, Deoxynivalenol 3-glucoside solution, analytical standard, Supelco®
Lieferant: Merck
32911-1ML
32911-1MLEA 574 EUR
32911-1ML
Organic reference standard, Deoxynivalenol 3-glucoside solution, analytical standard, Supelco®
Standards Organische Standards
Deoxynivalenol 3-glucoside solution is the major metabolite of deoxynivalenol (DON) and a modified mycotoxin. The conjugation of deoxynivalenol (DON) with glucose formed by the metabolism of plants results in the formation of deoxynivalenol-3-glucoside.
Deoxynivalenol 3-glucoside may be used as a reference standard for the determination of deoxynivalenol 3-glucoside in wheat and barley by liquid chromatography coupled to mass spectrometry.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=841d964bc10c155468e0502ba6c13ed784f01947&vid=2a3e9562bd2cb7b4fc877037fccf7cd9d23f67e7&prid=web&clid=SAPDAM