Organic reference standard, Deoxynivalenol 3-glucoside solution, analytical standard, Supelco®
Lieferant: Merck
32911-1ML
32911-1MLEA
574
EUR
32911-1ML
Organic reference standard, Deoxynivalenol 3-glucoside solution, analytical standard, Supelco®
Standards
Organische Standards
Deoxynivalenol 3-glucoside solution is the major metabolite of deoxynivalenol (DON) and a modified mycotoxin. The conjugation of deoxynivalenol (DON) with glucose formed by the metabolism of plants results in the formation of deoxynivalenol-3-glucoside.
Deoxynivalenol 3-glucoside may be used as a reference standard for the determination of deoxynivalenol 3-glucoside in wheat and barley by liquid chromatography coupled to mass spectrometry.