Organic reference standard, Neosolaniol, Supelco®
Lieferant: Merck
32932-5MG
32932-5MGEA 842 EUR
32932-5MG
Organic reference standard, Neosolaniol, Supelco®
Standards Organische Standards
Neosolaniol belongs to the class of type A trichothecene mycotoxins produced by various Fusarium molds.
Neosolaniol may be used as an analytical reference standard for the determination of the analyte in: Wheat samples by acetonitrile-based extraction and gas chromatography-tandem mass spectrometry (GC-MS/MS) as well as capillary GC with flame ionization detection and Cereals by acetonitrile-based extraction and GC coupled to mass spectrometry (MS).
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c9b7b7d89f858f8faa69449222e4f9771a4e01f&vid=5c9b7b7d89f858f8faa69449222e4f9771a4e01f&prid=web&clid=SAPDAM