Organic reference standard, Fumonisin B1, Supelco®
Lieferant: Merck
32936-5MG
32936-5MGEA
1100
EUR
32936-5MG
Organic reference standard, Fumonisin B1, Supelco®
Standards
Organische Standards
Fumonisins is a family of mycotoxins characterised by structurally related homologs. They are generally produced by fungi of Fusarium species.
It is used as an analytical reference standard for the determination of the analyte in swine liver by high performance liquid chromatography-fluorescence detection (HPLC-FLD) and in cereals by HPLC. It is a fungal metabolite believed to cause leukoencephalomalacia in horses.