4-Nitrophthalonitril 99%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

330590-5G
330590-5GEA 567 EUR
330590-5G
4-Nitrophthalonitril 99%, Sigma-Aldrich®
4-Nitrophthalonitril
Formel: C₈H₃N₃O₂
Schmelzpunkt: 144 °C
Lagertemperatur: Room Temperature
MDL: MFCD00040301
CAS-Nummer: 31643-49-9
EINECS: 250-748-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d5017611547c0142888b031a6fabf6680c9a5bf3&vid=9e18803c58c0ce96f4c16c6e0d1963e11c2287a2&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) White to Tan
Appearance (Form) Conforms to Requirements
Powder, Crystals, Crystalline Powder and/or Chunks
Proton NMR spectra Conforms to Structure
Carbon 54.7 - 56.3%
Nitrogen 23.9 - 24.6%
Purity (HPLC) ≥98.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR