1-Butanol ≥99,5% (durch GC), Puriss. p.a. Reag. Ph. Eur., ACS, ISO, Riedel-de Haën™

Lieferant: Honeywell Chemicals
Gefahr

Synonyme: 1-Butylalkohol, 1-Hydroxybutan, n-Butanol, Propylcarbinol

33065-45L 33065-1L 33065-2.5L 33065-30KG 33065-4X2.5L 33065-6X1L
33065-45L-REA 4244.1 EUR
33065-45L-R 33065-1L 33065-2.5L 33065-30KG 33065-4X2.5L 33065-6X1L
1-Butanol ≥99,5% (durch GC), Puriss. p.a. Reag. Ph. Eur., ACS, ISO, Riedel-de Haën™
1-Butanol

ACS GENERAL USE SOLVENTS


  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • Suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested to meet ACS general use specifications


Beilstein Number: 969148

Formel: H₃C(CH₂)₂CH₂OH
Siedepunkt: 119 °C (1,013 hPa)
Schmelzpunkt: <-90 °C
Dichte: 0,81 g/cm³ (20 °C)
Flammpunkt: 9,4 %(V)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002964
CAS-Nummer: 71-36-3
UN: 1120
ADR: 3,III
REACH: 01-2119484630-38
Merck Index: 13,01539
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9395eeb6150a74222da52460f53cedeccd93442&vid=fd8352a556914c5be7213c576355a88e130d1a7b&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (GC) Min. 99.5 %
Boiling range 116 - 118 °C
Refractive Index n20/D 1.399 (lit.)
Density (D 20/20) 0.808 - 0.812
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.1 %
Free acid (as C₃H₇COOH) Max. 0.005 %
Aluminium (Al) Max. 0.00005 %
Boron (B) Max. 0.000002 %
Barium (Ba) Max. 0.00001 %
Calcium (Ca) Max. 0.00005 %
Cadmium (Cd) Max. 0.000005 %
Cobalt (Co) Max. 0.000002 %
Chromium (Cr) Max. 0.000002 %
Copper (Cu) Max. 0.000002 %
Iron (Fe) Max. 0.00001 %
Magnesium (Mg) Max. 0.00001 %
Manganese (Mn) Max. 0.000002 %
Nickel (Ni) Max. 0.000002 %
Lead (Pb) Max. 0.00001 %
Tin (Sn) Max. 0.00001 %
Zinc (Zn) Max. 0.00001 %
iso-butanol (GC) Max. 0.15 %
n-Butyraldehyde (GC) Max. 0.01 %
Di-n-butyl ether (GC) Max. 0.1 %
2-Butanol (GC) Max. 0.05 %
Carbonyl compounds (as CO) Max. 0.01 %
APHA Max. 10


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR