Sicherheitssiegel, SampleSafe
Lieferant: BURKLE
5399-2902
331-0057EA 86.55 EUR
331-0057
Sicherheitssiegel, SampleSafe
Versiegeln Sicherheits-Siegel
SampleSafe seals are made of PP and are uniquely identified by a 9-digit serial number.
  • Serial numbering for clear identification
  • Stainless steel safety spring provides tamper protection
The seal ring (Ø 2 mm) is able to fit through small eyelets and slits. The tape is secured by a stainless steel spring with one-way tightening mechanism, which ensures a tamper-evident seal.