Combustion trays, ceramic, AL24
Lieferant: Thermo Fisher Scientific
33187.KF
33187.KFEA 416 EUR
33187.KF
Combustion trays, ceramic, AL24
Schiffchen Verbrennungsschiffchen
Pure Al₂O₃ Aluminium oxide AL24.
Slightly porous, good resistance to thermal shock, extremely good creep strength.