Aluminiumoxid Zement ≥95% (SiO₂ ∼ 5%)

Lieferant: Thermo Fisher Scientific

Synonyme: Aluminium(III) oxide, Alumina C, Aluminum oxide

33201.A1
33201.A1EA 105 EUR
33201.A1
Aluminiumoxid Zement ≥95% (SiO₂ ∼ 5%)
Aluminium oxide

CAS No.: 1344-28-1 Molecular Formula: Al2O3 Physical Form: Agglomerates 150 micron Odour: Odourless MDL No.: MFCD00211795

Formel: Al₂O₃
Molecular Weight: 101.96 g/mol
Siedepunkt: 2980
Schmelzpunkt: 2045
Dichte: 3.94 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003424
CAS-Nummer: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 13,00355

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR