Essigsäure ≥99,8%, Puriss. p.a. Reag. Ph. Eur., ACS, ISO, Fluka™
Lieferant: Honeywell Chemicals
|
Gefahr
|
Synonyme:
Ethansäure
33209-1L
33209-1L-GL
33209-2.5L
33209-2.5L-GL
33209-30KG
33209-4X2.5L
33209-4X2.5LGL
33209-60KG
33209-6X1L
33209-5L
33209-200L
33209-1LEA
65.11
EUR
33209-1L
33209-1L-GL
33209-2.5L
33209-2.5L-GL
33209-30KG
33209-4X2.5L
33209-4X2.5LGL
33209-60KG
33209-6X1L
33209-5L
33209-200L
Essigsäure ≥99,8%, Puriss. p.a. Reag. Ph. Eur., ACS, ISO, Fluka™
Essigsäure
Formel:
H₃CCOOH Molecular Weight: 60.05 g/mol Siedepunkt: 118 °C (1.013 hPa) Schmelzpunkt: 16 °C Dichte: 1.05 g/cm³ (20 °C) Flammpunkt: 40 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00036152 CAS-Nummer: 64-19-7 EINECS: 200-580-7 UN: 2789 ADR: 8 II REACH: 01-2119475328-30 |
|
Spezifikation Testergebnisse
Assay | 99.8 - 100.5 % |
Assay (GC) | Min. 99.8 % |
Density (D 20/20) | 1.052 - 1.053 |
Boiling range | 117 - 119 °C |
Congealing - Freezing Point | Min. 16.3 °C |
Non-volatile matter | Max. 0.0005 % |
Water (Karl Fischer) | Max. 0.4 % |
Silver (Ag) | Max. 0.000001 % |
Aluminium (Al) | Max. 0.000005 % |
Arsenic (As) | Max. 0.000001 % |
Barium (Ba) | Max. 0.000001 % |
Beryllium (Be) | Max. 0.000001 % |
Bismuth (Bi) | Max. 0.00001 % |
Calcium (Ca) | Max. 0.00002 % |
Cadmium (Cd) | Max. 0.000002 % |
Cobalt (Co) | Max. 0.000001 % |
Chromium (Cr) | Max. 0.000005 % |
Copper (Cu) | Max. 0.000001 % |
Iron (Fe) | Max. 0.00002 % |
Germanium (Ge) | Max. 0.000005 % |
Potassium (K) | Max. 0.00001 % |
Lithium (Li) | Max. 0.000001 % |
Magnesium (Mg) | Max. 0.00001 % |
Manganese (Mn) | Max. 0.000001 % |
Molybdenum (Mo) | Max. 0.000002 % |
Sodium (Na) | Max. 0.00005 % |
Nickel (Ni) | Max. 0.000005 % |
Lead (Pb) | Max. 0.000002 % |
Strontium (Sr) | Max. 0.000001 % |
Titanium (Ti) | Max. 0.000001 % |
Thallium (Tl) | Max. 0.000005 % |
Vanadium (V) | Max. 0.000001 % |
Zinc (Zn) | Max. 0.000005 % |
Zirconium (Zr) | Max. 0.00001 % |
Heavy metals (as Pb) | Max. 0.00005 % |
Chloride (Cl) | Max. 0.00005 % |
Phosphate (PO₄) | Max. 0.00005 % |
Sulfate (SO₄) | Max. 0.00005 % |
Acetaldehyde | Max. 0.0002 % |
Acetic aNHydride (GC) | Max. 0.01 % |
Mixable with H₂O | complying |
Titrable Base | Max. 0.0004 meq/g |
APHA | Max. 10 |
Appearance of the substance | complying |
K₂Cr₂O₇ reducing matter | complying |
KMnO₄ reducing substances | complying |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...