Ethylacetat ≥99,5% (durch GC), Puriss. p.a. Reag. Ph. Eur., ACS, ISO zur Analyse, Riedel-de Haën™

Lieferant: Honeywell Chemicals
Gefahr

Synonyme: Essigester, Essigsäureethylester, EtOAc

33211-7L 33211-18L 33211-45L 33211-180KG 33211-1L 33211-2.5L 33211-25L 33211-4X2.5L 33211-6X1L 33211-1000L
33211-7LEA 441.91 EUR
33211-7L 33211-18L-R 33211-45L-R 33211-180KG 33211-1L 33211-2.5L 33211-25L 33211-4X2.5L 33211-6X1L HONC33211-1000L
Ethylacetat ≥99,5% (durch GC), Puriss. p.a. Reag. Ph. Eur., ACS, ISO zur Analyse, Riedel-de Haën™
Ethylacetat

ACS GENERAL USE SOLVENTS


  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • Suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested to meet ACS general use specifications


Beilstein Number: 506104
RTECS Number: AH5425000
UNSPSC Code: 12191602

Formel: CH₃COOC₂H₅
Molecular Weight: 88.11 g/mol
Siedepunkt: 77 °C (1.013 hPa)
Schmelzpunkt: –84 °C
Dichte: 0.901 g/cm³
Flammpunkt: 11.5 %(V)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009171
CAS-Nummer: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3 II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (GC) Min. 99.5 %
Boiling range 76 - 78 °C
Density (D 20/4) 0.899 - 0.901
Refractive index (n 20/D) 1.3710 - 1.3730
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.05 %
Free acid (as CH₃COOH) Max. 0.0045 %
Aluminium (Al) Max. 0.00005 %
Boron (B) Max. 0.000002 %
Barium (Ba) Max. 0.00001 %
Bismuth (Bi) Max. 0.00001 %
Calcium (Ca) Max. 0.00005 %
Cadmium (Cd) Max. 0.000005 %
Cobalt (Co) Max. 0.000002 %
Chromium (Cr) Max. 0.000002 %
Copper (Cu) Max. 0.000002 %
Iron (Fe) Max. 0.00001 %
Potassium (K) Max. 0.00005 %
Lithium (Li) Max. 0.00001 %
Magnesium (Mg) Max. 0.00001 %
Molybdenum (Mo) Max. 0.00001 %
Manganese (Mn) Max. 0.000002 %
Sodium (Na) Max. 0.00005 %
Nickel (Ni) Max. 0.000002 %
Lead (Pb) Max. 0.00001 %
Tin (Sn) Max. 0.00001 %
Strontium (Sr) Max. 0.00001 %
Zinc (Zn) Max. 0.00001 %
methyl acetate Max. 0.1 %
Ethanol (GC) Max. 0.1 %
Isobutyl alcohol (GC) Max. 0.1 %
Methanol (GC) Max. 0.1 %
Reaction against H₂SO₄ complying
APHA Max. 10


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR