Ethylacetat 99,5 GC_ASSAY_METHOD, Puriss. p.a. Reag. Ph. Eur., ACS, ISO analytical reagent, Riedel-de Haën™
Lieferant: Honeywell Chemicals
|
Gefahr
|
Synonyme:
Essigester, Essigsäureethylester, EtOAc
33211-7L
33211-18L
33211-45L
33211-180KG
33211-1L
33211-2.5L
33211-25L
33211-4X2.5L
33211-6X1L
33211-1000L
33211-7LEA
441.91
EUR
33211-7L
33211-18L-R
33211-45L-R
33211-180KG
33211-1L
33211-2.5L
33211-25L
33211-4X2.5L
33211-6X1L
HONC33211-1000L
Ethylacetat 99,5 GC_ASSAY_METHOD, Puriss. p.a. Reag. Ph. Eur., ACS, ISO analytical reagent, Riedel-de Haën™
Ethylacetat
ACS GENERAL USE SOLVENTS
- Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- Suitable for less demanding applications such as routine isocratic HPLC
- Variations and decrease issues traceable to solvent contamination
- All solvents are tested to meet ACS general use specifications
Beilstein Number: 506104
RTECS Number: AH5425000
UNSPSC Code: 12191602
Formel:
CH₃COOC₂H₅ Molecular Weight: 88.11 g/mol Siedepunkt: 77 °C (1.013 hPa) Schmelzpunkt: –84 °C Dichte: 0.901 g/cm³ Flammpunkt: 11.5 %(V) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009171 CAS-Nummer: 141-78-6 EINECS: 205-500-4 UN: 1173 ADR: 3 II REACH: 01-2119475103-46 Merck Index: 13,03792 |
|
Spezifikation Testergebnisse
Assay (GC) | Min. 99.5 % |
Boiling range | 76 - 78 °C |
Density (D 20/4) | 0.899 - 0.901 |
Refractive index (n 20/D) | 1.3710 - 1.3730 |
Non-volatile matter | Max. 0.001 % |
Water (Karl Fischer) | Max. 0.05 % |
Free acid (as CH₃COOH) | Max. 0.0045 % |
Aluminium (Al) | Max. 0.00005 % |
Boron (B) | Max. 0.000002 % |
Barium (Ba) | Max. 0.00001 % |
Bismuth (Bi) | Max. 0.00001 % |
Calcium (Ca) | Max. 0.00005 % |
Cadmium (Cd) | Max. 0.000005 % |
Cobalt (Co) | Max. 0.000002 % |
Chromium (Cr) | Max. 0.000002 % |
Copper (Cu) | Max. 0.000002 % |
Iron (Fe) | Max. 0.00001 % |
Potassium (K) | Max. 0.00005 % |
Lithium (Li) | Max. 0.00001 % |
Magnesium (Mg) | Max. 0.00001 % |
Molybdenum (Mo) | Max. 0.00001 % |
Manganese (Mn) | Max. 0.000002 % |
Sodium (Na) | Max. 0.00005 % |
Nickel (Ni) | Max. 0.000002 % |
Lead (Pb) | Max. 0.00001 % |
Tin (Sn) | Max. 0.00001 % |
Strontium (Sr) | Max. 0.00001 % |
Zinc (Zn) | Max. 0.00001 % |
methyl acetate | Max. 0.1 % |
Ethanol (GC) | Max. 0.1 % |
Isobutyl alcohol (GC) | Max. 0.1 % |
Methanol (GC) | Max. 0.1 % |
Reaction against H₂SO₄ | complying |
APHA | Max. 10 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...