di-Ammoniumhydrogencitrat ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
33221.22
33221.36
33221.A3
33221.22EA
37.6
EUR
33221.22
33221.36
33221.A3
di-Ammoniumhydrogencitrat ≥99%
di-Ammoniumhydrogencitrat
Purity: 99+%
CAS No.: 3012-65-5
Molecular Formula: C6H14N2O7
Formula Weight: 226.19
Physical Form: Granular
Density: 1.48
Appearance: White
Odour: Odourless
Merck Reference: 14,512
MDL No.: MFCD00013068
Formel:
HOC(CO₂H)(CH₂CO₂NH₄)₂ Dichte: 1,48 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013068 CAS-Nummer: 3012-65-5 EINECS: 221-146-3 Merck Index: 14,512 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...