Cadmium(II)oxid ≥98,9%

Lieferant: Thermo Fisher Scientific
Gefahr

33235.30 33235.A1
33235.30EA 43.3 EUR
33235.30 33235.A1
Cadmium(II)oxid ≥98,9%
Cadmium(II)oxid

Purity: 98.9% CAS No.: 1306-19-0 Molecular Formula: CdO Formula Weight: 128.40 Physical Form: Powder Density: 8.15 Appearance: Red-brown Odour: Odourless Melting Point: 1430° Solubility: Practically insoluble in water. Soluble in dilute acids, slowly soluble in ammonium salts Merck Reference: 14,1623 UN No.: 2570 MDL No.: MFCD00010921

Formel: CdO
Siedepunkt: 1559 °C (1013 hPa)
Schmelzpunkt: 1430 °C
Dichte: 8,15
Lagertemperatur: Raumtemperatur
MDL: MFCD00010921
CAS-Nummer: 1306-19-0
EINECS: 215-146-2
UN: 2570
ADR: 6.1,III
Merck Index: 14,1623

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR