Essigsäure (Eisessig) ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99+%
CAS No.: 64-19-7
Molecular Formula: CH3CO2H
Formula Weight: 60.05
Flash Point: 40°(104°F)
Physical Form: Liquid
Density: 1.049
Refractive Index: 1.3721
Odour: Pungent
pH: 2,5
Melting Point: 16.6°
Boiling Point: 118.1°
Merck Reference: 14,55
UN No.: 2789
MDL No.: MFCD00036152
Formel:
H₃CCOOH Siedepunkt: 118,1 °C Dichte: 1,049 Flammpunkt: 40 °C (104 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00036152 CAS-Nummer: 64-19-7 EINECS: 200-580-7 UN: 2789 ADR: 8,II Merck Index: 14,55 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...