Ammoniaklösung 28,0 - 30,0% ACS
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Ammoniumhydroxid, Ammoniak
33285.30
33285.A1
33285.D9
33285.30EA
34.1
EUR
33285.30
33285.A1
33285.D9
Ammoniaklösung 28,0 - 30,0% ACS
Ammoniaklösung
CAS No.: 1336-21-6
Molecular Formula: NH4OH
Formula Weight: 35.05
Physical Form: Liquid
Density: 0.90
Appearance: Colourless
Odour: Ammonia-like
pH: 12
Merck Reference: 14,494
UN No.: 2672
MDL No.: MFCD00066650
Formel:
NH₃ Siedepunkt: 32…37,7 °C (1013 hPa) Schmelzpunkt: –60…–57,5 °C Dichte: 0,90 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066650 CAS-Nummer: 1336-21-6 EINECS: 215-647-6 UN: 2672 ADR: 8,III Merck Index: 14,494 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...