Calciumsulfat Dihydrat ≥99%
Lieferant: Thermo Fisher Scientific
Synonyme:
Pflaster Paris
33301.A1
33301.A7
33301.A1EA
78.3
EUR
33301.A1
33301.A7
Calciumsulfat Dihydrat ≥99%
Calciumsulfat Dihydrat
Purity: 99%
CAS No.: 10101-41-4
Molecular Formula: CaSO4.2H2O
Formula Weight: 172.17 (136.14anhy)
Physical Form: Powder
Density: 2.32
Melting Point: 128° -1.5H2O
Boiling Point: 163° -2H2O
Merck Reference: 14,1706
MDL No.: MFCD00149625
Formel:
CaSO₄·2H₂O Siedepunkt: 163 °C (-2H₂O) Schmelzpunkt: 128 °C (-1,5H₂O) Dichte: 2,32 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149625 CAS-Nummer: 10101-41-4 EINECS: 231-900-3 Merck Index: 14,1706 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...