Cyclohexanon 99+% ACS
Lieferant: Thermo Fisher Scientific
|
Achtung
|
33309.AP
33309.K2
33309.K7
33309.M1
33309.APEA
45.3
EUR
33309.AP
33309.K2
33309.K7
33309.M1
Cyclohexanon 99+% ACS
Cyclohexanone
Purity: 99+%
CAS No.: 108-94-1
Molecular Formula: C6H10O
Formula Weight: 98.15
Flash Point: 47°(116°F)
Physical Form: Liquid
Density: 0.9478
Refractive Index: 1.4507
Melting Point: -26°
Boiling Point: 154-156°
Beilstein Reference: 385735
Merck Reference: 14,2726
UN No.: 1915
MDL No.: MFCD00001625
Formel:
C₆H₁₀(=O) Siedepunkt: 154…156 °C Schmelzpunkt: –26 °C Dichte: 0,9478 Flammpunkt: 47 °C (116 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001625 CAS-Nummer: 108-94-1 EINECS: 203-631-1 UN: 1915 ADR: 3,III REACH: 01-2119453616-35 Merck Index: 14,2726 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...