Blei(II)carbonat basisch ACS

Lieferant: Thermo Fisher Scientific
Gefahr

33328.30
33328.30EA 161 EUR
33328.30
Blei(II)carbonat basisch ACS
Blei(II)carbonat basisch

CAS No.: 1319-46-6 Molecular Formula: C2H2O8Pb3 Formula Weight: 775.63 Physical Form: Powder Density: 6.14 Appearance: White Odour: Odourless Melting Point: 400° dec. Solubility: Soluble in acids, insoluble in water or alcohol Merck Reference: 14,1005 UN No.: 3077 MDL No.: MFCD00078155


Ceramic glaze, exterior paint

Formel: (PbCO₃)₂·Pb(OH)₂
Dichte: 6,14
Lagertemperatur: Raumtemperatur
MDL: MFCD00078155
CAS-Nummer: 1319-46-6
EINECS: 215-290-6
UN: 3077
ADR: 9,III
Merck Index: 14,1005

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR