Blei(II)oxid silberarm

Lieferant: Thermo Fisher Scientific
Gefahr

33330.36 33330.A3
33330.36EA 32.4 EUR
33330.36 33330.A3
Blei(II)oxid silberarm
Blei(II)oxid

CAS No.: 1317-36-8 Molecular Formula: PbO Formula Weight: 223.19 Physical Form: Powder Density: 9.53 Odour: Odourless Merck Reference: 14,5413 UN No.: 3077 MDL No.: MFCD00011164


As storage batteries, ceramic cements and fluxes, pottery and glazes, in chromium pigments, in oil refining, in paints, and in the assay of precious metal ores

Formel: PbO
Siedepunkt: 1470 °C (1013 hPa)
Schmelzpunkt: 886 °C
Dichte: 9,53
Lagertemperatur: Raumtemperatur
MDL: MFCD00011164
CAS-Nummer: 1317-36-8
EINECS: 215-267-0
UN: 3077
ADR: 9,III
Merck Index: 14,5413

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR