Mangan(II)nitrat 50% (w/w) in wässriger Lösung
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Manganese dinitrate, Manganous nitrate
33340.36
33340.A3
33340.36EA
58.2
EUR
33340.36
33340.A3
Mangan(II)nitrat 50% (w/w) in wässriger Lösung
Mangan(II)nitrat
CAS No.: 10377-66-9
Molecular Formula: MnN2O6
Formula Weight: 178.95
Physical Form: Liquid
Appearance: Pink
Odour: Amine-like
pH: 1.0
UN No.: 3093
MDL No.: MFCD00011112
Formel:
Mn(NO₃)₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011112 CAS-Nummer: 10377-66-9 EINECS: 233-828-8 UN: 3093 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...