α-Naphtholphthalein Indikator, Fluka™

Lieferant: Honeywell Chemicals
Achtung

Synonyme: 3,3-Bis(4-hydroxynaphthalen-1-yl)isobenzofuran-1(3H)-one

33424-10G
33424-10GEA 532.32 EUR
33424-10G
α-Naphtholphthalein Indikator, Fluka™
α-Naphtholphthalein
Formel: C₂₈H₁₈O₄
Molecular Weight: 418,45 g/mol
Schmelzpunkt: 238-240 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00036202
CAS-Nummer: 596-01-0
EINECS: 209-875-5

Bestellen

Spezifikation Testergebnisse

Change range (pH 7.1-8.3) complying
Test of sensitivity complying

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR