, pellets, Ø 1.6 mm, Fluka™
Lieferant: Honeywell Chemicals
Synonyme:
Molekularsieb A3
334286-1KG
334286-1KGEA
232.84
EUR
334286-1KG
, pellets, Ø 1.6 mm, Fluka™
Molekularsieb 3A (0,3 nm, 3 Å)
Formel:
KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O Schmelzpunkt: 400 °C Dichte: 2 g/cm³ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00147627 CAS-Nummer: 1318-02-1 |
Spezifikation Testergebnisse
Particle Size | 1.6 mm |
Form | pellets |
Granulation | complying |
Pore Size (Ång Ström) | 3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...