o-Dianisidin, Supelco®

Lieferant: Merck

Synonyme: 3,3'-Dimethoxybenzidin

M-Clarity™ Qualitätsstufe = MQ100

33430-10G
33430-10GEA 112 EUR
33430-10G
o-Dianisidin, Supelco®
o-Dianisidin
Formel: C₁₄H₁₆N₂O₂
Molecular Weight: 244,29 g/mol
MDL: MFCD00008372
CAS-Nummer: 119-90-4
EINECS: 204-355-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6fe4e2fe81a1e4bf30b85a086aa2fb02476a07c1&vid=a27ef7455277a137333b84f01d437a7fb56d7ac5&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR