Molekularsieb A5 (0,5 nm, 5 Å), Plätzchen, Ø 1.6 mm, Fluka™
Lieferant: Honeywell Chemicals
Synonyme:
Linde 5A, Molecular sieves, 5A 3.0mm-5.0mm pellets
334316-1KG
334316-1KGEA
242.23
EUR
334316-1KG
Molekularsieb A5 (0,5 nm, 5 Å), Plätzchen, Ø 1.6 mm, Fluka™
Molekularsieb A5 (0,5 nm, 5 Å)
MDL:
MFCD00131613 CAS-Nummer: 69912-79-4 |
Spezifikation Testergebnisse
Particle Size | 1.6 mm |
Form | pellets |
Granulation | complying |
Pore Size (Ång Ström) | 5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...