Ethylenglycolmonomethylether (2-Methoxyethanol) ≥99,5% (durch GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™

Lieferant: Honeywell Chemicals
Gefahr

Synonyme: Methylglycol, Ethylenglycolmonomethylether

33457-2.5L
33457-2.5LEA 177.75 EUR
33457-2.5L
Ethylenglycolmonomethylether (2-Methoxyethanol) ≥99,5% (durch GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Ethylenglycolmonomethylether (2-Methoxyethanol)

ACS GENERAL USE SOLVENTS


  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • Suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested to meet ACS general use specifications

Formel: C₃H₈O₂
Molecular Weight: 76,1 g/mol
Siedepunkt: 124 °C (1,013 hPa)
Schmelzpunkt: –85 °C
Dichte: 0,97 g/cm³ (20 °C)
Flammpunkt: 38 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002867
CAS-Nummer: 109-86-4
EINECS: 203-713-7
UN: 1188
ADR: 3,III
Merck Index: 12,06118

Bestellen

Spezifikation Testergebnisse

Assay (GC) Min. 99.5 %
Boiling range 123 - 126 °C
Explosion Limit 24.5 %
Density (D 20/20) 0.966 - 0.968
Refractive index (n 20/D) 1.4010 - 1.4030
Non-volatile matter Max. 0.002 %
Water (Karl Fischer) Max. 0.1 %
Titrable Acid Max. 0.002 meq/g
Aluminium (Al) Max. 0.00005 %
Boron (B) Max. 0.000002 %
Barium (Ba) Max. 0.00001 %
Calcium (Ca) Max. 0.00005 %
Cadmium (Cd) Max. 0.000005 %
Cobalt (Co) Max. 0.000002 %
Chromium (Cr) Max. 0.000002 %
Iron (Fe) Max. 0.00004 %
Magnesium (Mg) Max. 0.00001 %
Manganese (Mn) Max. 0.000002 %
Nickel (Ni) Max. 0.000002 %
Lead (Pb) Max. 0.00001 %
Tin (Sn) Max. 0.00001 %
Zinc (Zn) Max. 0.0004 %
Acetaldehyde Max. 0.001 %
Formaldehyde Max. 0.001 %
Peroxides (as H₂O₂) Max. 0.0003 %
APHA Max. 10

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR