4,4′-Dinitro-2-biphenylamine ≥98%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: 4,4'-Dinitro-1,1'-biphenyl-2-amine

335428-25G
335428-25GEA 269 EUR
335428-25G
4,4′-Dinitro-2-biphenylamine ≥98%, Sigma-Aldrich®
4,4′-Dinitro-2-biphenylamine

4,4'-Dinitro-2-biphenylamine,98%

Formel: C₁₂H₉N₃O₄
Molecular Weight: 259,22 g/mol
Schmelzpunkt: 208…210 °C (Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00075056
CAS-Nummer: 51787-75-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a352b865b64a970475cf89cffe650403e698a3cd&vid=f4e8895337932954f9acbf35a2f9430eaeccc931&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR