Tris(4-nitrophenyl)amine, technische Qualität, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ100
335495-5G
335495-5GEA
105
EUR
335495-5G
Tris(4-nitrophenyl)amine, technische Qualität, Sigma-Aldrich®
Tris(4-nitrophenyl)amine
Formel:
C₁₈H₁₂N₄O₆ Schmelzpunkt: >300 °C (Lit.) Lagertemperatur: Room Temperature |
MDL:
MFCD00024636 CAS-Nummer: 20440-93-1 |
Spezifikation Testergebnisse
Appearance | Yellow-green or gold powder |
Infrared spectrum | Conforms to structure |
Proton NMR spectrum | Conforms to structure |
High pressure liquid | Typically ca. 97% |
Elemental analysis | Carbon typically 54.8% - 58.8%, Nitrogen typically 14.2% - 15.2% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...