Alcian Blue, pyridine variant ≥85 % (Farbstoffgehalt), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: Alcian Blue-tetrakis(methylpyridinium) chloride

335630-25G
335630-25GEA 312 EUR
335630-25G
Alcian Blue, pyridine variant ≥85 % (Farbstoffgehalt), Sigma-Aldrich®
Alcian Blue, pyridine variant

Alcian Blue, pyridine variant, Dye content ≥85 % (Alcian Blue-tetrakis(methylpyridinium) chloride)

Formel: C₅₆H₄Cl₄CuN₁₂
Molecular Weight: 1086,36 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 123439-83-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ad318019658248e6e82d773282bcc84d41dc4256&vid=47ab307c25cf9d075ceb9858e37949da4bcae40b&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR