Iron(III) meso-tetraphenylporphine-µ-oxo dimer

Lieferant: Thermo Scientific

Synonyme: Iron(III) meso-tetraphenylporphine-mu-oxo dimer, Meso-5,10,15,20-Tetraphenyl-21H,23H-porphineironu-oxodimer

33738.ME
33738.MEEA 254 EUR
33738.ME
Iron(III) meso-tetraphenylporphine-µ-oxo dimer
Iron(III) meso-tetraphenylporphine-µ-oxo dimer

CAS No.: 12582-61-5 Molecular Formula: C88H56Fe2N8O Formula Weight: 1353.16 Physical Form: Solid MDL No.: MFCD00058898

Formel: C₈₈H₅₆Fe₂N₈O
Molecular Weight: 1353,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00058898
CAS-Nummer: 12582-61-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR