Hydroxynaphtholblau Indicator_Metal_titration, Fluka™

Lieferant: Honeywell Chemicals

Synonyme: 1-(2-Hydroxy-4-sulpho-1-naphthylazo)-2-naphthol-3,6-disulphonic acid trisodium salt

33936-10G 33936-50G
33936-10GEA 106.97 EUR
33936-10G 33936-50G
Hydroxynaphtholblau Indicator_Metal_titration, Fluka™
Hydroxynaphthol blue
Formel: C₂₀H₁₁N₂Na₃O₁₁S₃
Molecular Weight: 620.47 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00036393
CAS-Nummer: 63451-35-4
EINECS: 264-197-7

Bestellen

Spezifikation Testergebnisse

Identity (LC-MS) complying
Absorption maximum 640 - 650 nm
Absorbtion (1%/1cm) Min. 325
Suitability for Complexometry complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR