Kieselgel, BAKER ANALYZED® ACS (6-18 Mesh) trocknungsfördernd, mit Feuchtigkeitsindikator, J.T.Baker®

Lieferant: Avantor

3401-01EA 0 EUR
3401-01
Kieselgel, BAKER ANALYZED® ACS (6-18 Mesh) trocknungsfördernd, mit Feuchtigkeitsindikator, J.T.Baker®
Silica gel

6 to 18 mesh. Indicating (cobalt chloride). Lot analysis on label.


Color Change:
Blue.................Activated
Violet..........10% Moisture Absorbed
Pink............19% Moisture Absorbed
Pale Pink.......28% Moisture Absorbed

Formel: SiO₂
Siedepunkt: 2230 °C (1013 hPa)
Schmelzpunkt: 1710 °C
Dichte: 2,10 kg/m³
MDL: MFCD00011232
CAS-Nummer: 63231-67-4

Bestellen

Spezifikation Testergebnisse

Meets ACS Reagent Chemical Requirements ≤ 6.0 %
For Laboratory,Research,or Manufacturing Use ≥ 27 %
Loss on Heating at 200°C ≤ 6.0 %
Suitability for Moisture Absorption ≥ 27 %
Suitability as Indicating Desiccant Passes Test
Mesh - Thru U.S. No. 6 Sieve ≥ 90 %
Mesh - Thru U.S. No. 18 Sieve ≤ 10 %
Color Change: Blue.................Activated Violet..........10% Moisture Absorbed Pink............19% Moisture Absorbed Pale Pink.......28% Moisture Absorbed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR