2,4,6-Trichloranisol-[D5], Supelco®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

34023-50MG-R
34023-50MG-REA 598 EUR
34023-50MG-R
2,4,6-Trichloranisol-[D5], Supelco®
2,4,6-Trichloranisol-[D5]
Formel: C₇D₅Cl₃O
Lagertemperatur: Raumtemperatur
CAS-Nummer: 352439-08-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e12ef33eb2b42db4e960af9e2fe4c18a03ea5cbb&vid=3c27dec55f7a0f7df7503bb62839961139fc2a56&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR