Edelfosine ≥95% (durch TLC), weißer Feststoff, Millipore®

Lieferant: Merck Millipore Calbiochem

Synonyme: 1-O-Octadecyl-2-O-methyl-rac-glycero-3-phosphocholine

341207-5
341207-5EA 120 EUR
341207-5
Edelfosine ≥95% (durch TLC), weißer Feststoff, Millipore®
Edelfosine

A cell-permeable and reversible cytotoxic agent that shows selective cytocidal activity against neoplastic cells and virally transformed cells. Selectively inhibits phosphatidylinositol-specific phospholipase C but does not inhibit phosphatidylcholine-specific PLC or PLD. Promotes apoptosis in mitogen-activated, but not resting, T lymphocytes in a dose and time-dependent manner. It also promotes rapid and selective apoptosis in human leukemic cells.

Formel: C₂₇H₅₈NO₆P
Molecular Weight: 523.736 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00135190
CAS-Nummer: 70641-51-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR