Bovine (foetal) fetuin (from serum)

Lieferant: Merck Millipore Calbiochem

341506-1
341506-1EA 241 EUR
341506-1
Bovine (foetal) fetuin (from serum)
Proteine und Peptide

Fetuin extracted from foetal bovine serum. Fetuin is an α-globulin that inhibits trypsin activity and promotes cell attachment, growth and differentiation in many different culture systems.


  • Green freeze dried solid
  • Lyophilised from 0,5% (NH₄)₂SO₄
  • Purity ≥70% by electrophoresis


Fetuin also contains both N- and O-glycosidically-linked carbohydrates. It functions as an opsonin for cationic-deactivating molecules in macrophages.

Bestellen

TECHNISCHE DATEN

Protein/Peptid-Name Fetuin
Environmentally Preferable
Spezies Bovine (foetal)
Ursprung Serum
Purity >70% (by electrophoresis)
Zusammensetzung Lyophilised
Lagerbedingungen –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR