Titanium(IV) oxysulphate-Lösung zur Bestimmung von Wasserstoffperoxid (H₂O₂) according to DIN 38 409, part 15 DEV-18, Supelco®

Lieferant: Merck

Synonyme: Titanium(IV) oxide sulphate

M-Clarity™ Qualitätsstufe = MQ100

34244-1L
34244-1LEA 132 EUR
34244-1L
Titanium(IV) oxysulphate-Lösung zur Bestimmung von Wasserstoffperoxid (H₂O₂) according to DIN 38 409, part 15 DEV-18, Supelco®
Titanium(IV) oxysulphate

Titanium(IV) oxysulfate is known to form a yellow complex upon reaction with hydrogen peroxide (H2O2) which facilitates the latter′s determination by spectrophotometric methods.

Formel: TiOSO₄·aq
Molecular Weight: 159,93 g/mol (anhy)
MDL: MFCD00149873
CAS-Nummer: 13825-74-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=466af2a84b388da32b7e3754727a830bfce9ffc7&vid=56a15059a24b1c089131cf693a1e6e5c6733a2d5&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR