FITC (Fluorescein-5-isothiocyanat, Fluoresceinisothiocyanat Isomer I)
Lieferant: Merck Millipore (Oncogene)
|
Achtung
|
34321-1
34321-1EA
326
EUR
34321-1
FITC (Fluorescein-5-isothiocyanat, Fluoresceinisothiocyanat Isomer I)
FITC (Fluorescein-5-isothiocyanat, Fluoresceinisothiocyanat Isomer I)
Formel:
C₂₁H₁₁NO₅S Lagertemperatur: Kühlschrank |
MDL:
MFCD00005063 CAS-Nummer: 3326-32-7 EINECS: 222-042-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...