5-Fluor-2'-deoxyuridin (Floxuridin), Millipore®

Lieferant: Merck Millipore Calbiochem

Synonyme: 5-Fluoro-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 5-Fluoro-2'-deoxyuridine, Floxuridine

343333-50
343333-50EA 98.3 EUR
343333-50
5-Fluor-2'-deoxyuridin (Floxuridin), Millipore®
5-Fluor-2'-deoxyuridin (Floxuridin)
Formel: C₉H₁₁FN₂O₅
MDL: MFCD00006530
CAS-Nummer: 50-91-9
EINECS: 200-072-5
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR