Ammoniumnitrat ≥95,0%, Kristalle, AR® ACS, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials
Gefahr

3436-06EA 0 EUR
3436-06 3436-20
Ammoniumnitrat ≥95,0%, Kristalle, AR® ACS, Macron Fine Chemicals™
Ammoniumnitrat
Formel: NH₄NO₃
Siedepunkt: 230 °C (1013 hPa)
Schmelzpunkt: 169 °C
Dichte: 1,73 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011425
CAS-Nummer: 6484-52-2
EINECS: 229-347-8
UN: 1942
ADR: 5.1,III
Merck Index: 13,00538

Bestellen

Spezifikation Testergebnisse

Meets ACS Reagent Chemical Requirements = 95.0 %
For Laboratory,Research,or Manufacturing Use = 5 ppm
ACS - Assay (as NH₄NO₃) ≥ 95.0 %
ACS - Chloride (Cl) ≤ 5 ppm
ACS - Heavy Metals (as Pb) ≤ 5 ppm
ACS - Insoluble Matter ≤ 0.005 %
ACS - Iron (Fe) ≤ 2 ppm
ACS - Nitrite (NO₂) Passes Test
ACS - pH of 5% Solution at 25°C 4.5 - 6.0
ACS - Phosphate (PO₄) ≤ 5 ppm
ACS - Residue after Ignition ≤ 0.01 %
ACS - Sulfate (SO₄) ≤ 0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR