Diluid MINDRAY für die Hämatologie, J.T.Baker®
Lieferant: Avantor
3439.9020PCEA 34.17 EUR
3439.9020PC
Diluid MINDRAY für die Hämatologie, J.T.Baker®
Diluid
Bitte beachten In vitro diagnostic medical devices only for professional use.
IVDLogo
Bestellen
Spezifikation Testergebnisse
Background PLT (x10E9/L) max. 5
Background RBC (x10E12/L) max. 0.02
Background WBC (x10E9/L) max. 0.3
Conductivity (mS) 16.00 - 21.00
Osmolality (mOsm/kg) 325 - 335
Performance Test passes test
pH at 25°C 6.90 - 7.10
Total Bacterial Growth (CFU/100ml) max. 5
Osmolality is measured directly after production. In course of time the actual value will increase. This will not influence the performance of the reagent.