Diluid MINDRAY für die Hämatologie, J.T.Baker®
Lieferant: Avantor
3439.9020PCEA
34.17
EUR
3439.9020PC
Diluid MINDRAY für die Hämatologie, J.T.Baker®
Diluid
Bitte beachten In vitro diagnostic medical devices only for professional use.
![]() |
Spezifikation Testergebnisse
Background PLT (x10E9/L) | max. 5 |
Background RBC (x10E12/L) | max. 0.02 |
Background WBC (x10E9/L) | max. 0.3 |
Conductivity (mS) | 16.00 - 21.00 |
Osmolality (mOsm/kg) | 325 - 335 |
Performance Test | passes test |
pH at 25°C | 6.90 - 7.10 |
Total Bacterial Growth (CFU/100ml) | max. 5 |
Osmolality is measured directly after production. In course of time the actual value will increase. This will not influence the performance of the reagent. |