cFMS Receptor Inhibitor IV ≥95% (durch HPLC), Calbiochem®, Sigma-Aldrich®

Lieferant: Merck Millipore Calbiochem

Synonyme: 5-Cyano-N-(2,5-di(piperidin-1-yl)phenyl)furan-2-carboxamide, CSF-1 Receptor Inhibitor IV

M-Clarity™ Qualitätsstufe = MQ100

344039-5
344039-5EA 235 EUR
344039-5
cFMS Receptor Inhibitor IV ≥95% (durch HPLC), Calbiochem®, Sigma-Aldrich®
cFMS Receptor Inhibitor IV

The cFMS Receptor Inhibitor IV, also referenced under CAS 959626-45-0, controls the biological activity of cFMS. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.


General description
A cell-permeable furan carboxamide compound that acts as a potent, reversible, and ATP-competitive cFMS/CSF-1R inhibitor that blocks cFMS autophosphorylation both in CSF-1/M-CSF-stimulated HEK-293 cells (IC50 = 76 nM) and in cell-free in vitro kinase assays (IC50 = 17 nM).


Verpackung: Packaged under inert gas

Formel: C₂₂H₂₆N₄O₂
Molecular Weight: 378,47 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD10568148
CAS-Nummer: 959626-45-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR