2-Fluor-α-methyl-4-biphenylessigsäure, Calbiochem®

Lieferant: Merck Millipore Calbiochem

Synonyme: Ansaid, 2-(2-Fluoro-[1,1'-biphenyl]-4-yl)propanoic acid, Flurbiprofen, Froben, Antadys

M-Clarity™ Qualitätsstufe = MQ200

344079-100
344079-100EA 71.1 EUR
344079-100
2-Fluor-α-methyl-4-biphenylessigsäure, Calbiochem®
2-Fluor-α-methyl-4-biphenylessigsäure
Formel: C₁₅H₁₃FO₂
Molecular Weight: 244,265 g/mol
MDL: MFCD00079303
CAS-Nummer: 5104-49-4
EINECS: 225-827-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR