n-Hexan 95 GC_ASSAY_METHOD, CHROMASOLV™ Pest_Residual, dioxins_furans_PCB analysis, Riedel-de Haën™
Lieferant: Honeywell Chemicals
|
Gefahr
|
Synonyme:
Hexyl hydride, Hexanen, Hexane
34412-7L
34412-2.5L
34412-7LEA
485.88
EUR
34412-7L
34412-2.5L
n-Hexan 95 GC_ASSAY_METHOD, CHROMASOLV™ Pest_Residual, dioxins_furans_PCB analysis, Riedel-de Haën™
n-Hexan
- High purity chromatography solvents
- Suitability tested for specified use from HPLC up to UHPLC-MS applications.
Formel:
H₃C(CH₂)₄CH₃ Molecular Weight: 86.18 g/mol Siedepunkt: 66…69 °C(1.013 hPa) Schmelzpunkt: –95 °C Dichte: 0.660…0.668 g/cm³ (20 °C) Flammpunkt: 7.4 %(V) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009520 CAS-Nummer: 110-54-3 EINECS: 203-777-6 UN: 1208 ADR: 3 II Merck Index: 13,04712 |
|
Spezifikation Testergebnisse
Assay (GC) | Min. 95 % |
Non-volatile matter | Max. 0.0005 % |
Water (Karl Fischer) | Max. 0.01 % |
Suitability for GC-MS of dioxines, furanes, PCPs | complying |
Vapor Density | ~3 (vs air) |
Vapor Pressure | 5.2 psi ( 37.7 °C) |
Refractive Index | n20/D 1.375 (lit.) |
Explosion Limit | 7.7 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...