Dibutylether ≥99,5% (durch GC), Puriss. p.a. niedriger aromatische Kohlenwasserstoffe-Gehalt, Riedel-de Haën™
Lieferant: Honeywell Chemicals
|
Achtung
|
34462-1L
34462-1LEA
178.53
EUR
34462-1L
Dibutylether ≥99,5% (durch GC), Puriss. p.a. niedriger aromatische Kohlenwasserstoffe-Gehalt, Riedel-de Haën™
Dibutylether
Beilstein Number: 1732752
RTECS Number: EK5425000
UNSPSC Code: 12190000
Formel:
C₈H₁₈O Siedepunkt: 140 °C Schmelzpunkt: -95 °C Dichte: 0,77 g/cm³ Flammpunkt: 8,5 %(V) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009461 CAS-Nummer: 142-96-1 EINECS: 205-575-3 UN: 1149 ADR: 3,III Merck Index: 13,01568 |
Spezifikation Testergebnisse
Autoignition Temperature | 365 °F |
Assay (GC) | Min. 99.5 % |
Refractive index (n 20/D) | 1.398 - 1.400 |
Non-volatile matter | Max. 0.002 % |
Water (Karl Fischer) | Max. 0.05 % |
Aluminium (Al) | Max. 0.5 ppm |
Barium (Ba) | Max. 0.1 ppm |
Bismuth (Bi) | Max. 0.1 ppm |
Calcium (Ca) | Max. 0.5 ppm |
Cadmium (Cd) | Max. 0.05 ppm |
Cobalt (Co) | Max. 0.02 ppm |
Chromium (Cr) | Max. 0.02 ppm |
Copper (Cu) | Max. 0.02 ppm |
Iron (Fe) | Max. 0.1 ppm |
Potassium (K) | Max. 0.5 ppm |
Lithium (Li) | Max. 0.1 ppm |
Magnesium (Mg) | Max. 0.1 ppm |
Manganese (Mn) | Max. 0.02 ppm |
Molybdenum (Mo) | Max. 0.1 ppm |
Sodium (Na) | Max. 0.5 ppm |
Nickel (Ni) | Max. 0.02 ppm |
Lead (Pb) | Max. 0.1 ppm |
Strontium (Sr) | Max. 0.1 ppm |
Zinc (Zn) | Max. 0.1 ppm |
Peroxides (as H₂O₂) | Max. 20 ppm |
Aromatic compounds (as Xylene) | Max. 0.05 % |
Refractive Index | n20/D 1.399 (lit.) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...