D(+)-Galactose, Millipore®

Lieferant: Merck Millipore Calbiochem

Synonyme: (2R,3S,4S,5R)-Pentahydroxyhexanal

3455-100
3455-100EA 109 EUR
3455-100
D(+)-Galactose, Millipore®
D(+)-Galactose
Formel: C₆H₁₂O₆
Molecular Weight: 180,16 g/mol
Siedepunkt: 527 °C (1013 hPa)
Schmelzpunkt: 163…169 °C
Dichte: 1,5 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00151230
CAS-Nummer: 59-23-4
EINECS: 200-416-4
Merck Index: 13,04356
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=508b1d21a6801ddfb1a5f13a64cb06e6f95e8922&vid=3c02003b1d12c9bfd11c2e6faf5cc37d4325d208&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR