Natriumacetat Trihydrat 99,0-101,0%, Kristalle, BAKER ANALYZED® ACS, J.T.Baker®

Lieferant: Avantor
Achtung

3460-07EA 521 EUR
3460-07 3460-01 3460-R
Natriumacetat Trihydrat 99,0-101,0%, Kristalle, BAKER ANALYZED® ACS, J.T.Baker®
Natriumacetat Trihydrat
Formel: H₃CCOONa·3H₂O
Siedepunkt: 120…123 °C (1013 hPa)
Schmelzpunkt: 58 °C
Dichte: 1,45 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00071557
CAS-Nummer: 6131-90-4
EINECS: 204-823-8
Merck Index: 13,08642
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=313c1d45673fab192565649f8cbb7ddff555de39&vid=1ad37a966605626c95117735c83b16eaea381fbc&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Meets ACS Reagent Chemical Requirements 99.0 - 101.0%
For Laboratory,Research,or Manufacturing Use ≤0.005%
Assay (CH₃COONa·3H₂O) 99.0 - 101.0 %
Insoluble Matter ≤0.005 %
pH of 5% Solution at 25°C 7.5 - 9.2
Chloride (Cl) ≤0.001 %
Sulfate (SO₄) ≤0.002 %
Calcium (Ca) ≤0.005 %
ACS - Magnesium (Mg) ≤0.002 %
Substances Reducing Permanganate Passes Test
Potassium (K) ≤0.005 %
Phosphate (PO₄) ≤5 ppm
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR