Rinsing solution I, CHROMASOLV™ LabReady™, Lösungsmittelgemisch Spülmittel für LC-MS, Riedel-de Haën™

Lieferant: Honeywell Chemicals
Gefahr

34689-1L
34689-1LEA 65.16 EUR
34689-1L
Rinsing solution I, CHROMASOLV™ LabReady™, Lösungsmittelgemisch Spülmittel für LC-MS, Riedel-de Haën™
Rinsing solution I

  • High purity chromatography solvents
  • Suitability tested for specified use from HPLC up to UHPLC-MS applications.

Flammpunkt: 22 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011674
UN: 1219
ADR: 3,II

Bestellen

Spezifikation Testergebnisse

Assay of water 49 - 51 Vol.%
Identity 2-propanol (GC) complying
Non-volatile matter Max. 0.001 %
Silver (Ag) Max. 0.1 ppm
Aluminium (Al) Max. 0.5 ppm
Barium (Ba) Max. 0.1 ppm
Calcium (Ca) Max. 0.1 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.1 ppm
Potassium (K) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.02 ppm
Sodium (Na) Max. 0.1 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.1 ppm
Tin (Sn) Max. 0.1 ppm
Zinc (Zn) Max. 0.1 ppm
Organic impurities GC Max. 0.1 Fl.%
Absobance at 254 nm Max. 0.008
Absorbance at 230 nm Max. 0.045
Absorbance at 210 nm Max. 0.096
Suitability for the LC-MS complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR