n-Hexan ≥97,0% (durch GC), CHROMASOLV™ für die HPLC, Riedel-de Haën™
Lieferant: Honeywell Chemicals
|
Gefahr
|
34859-45L
34859-7L
34859-1L
34859-2.5L
34859-4X2.5L
34859-6X1L
34859-200L
34859-45LEA
6771.47
EUR
34859-45L
34859-7L
34859-1L
34859-2.5L
34859-4X2.5L
34859-6X1L
HONC34859-200L
n-Hexan ≥97,0% (durch GC), CHROMASOLV™ für die HPLC, Riedel-de Haën™
n-Hexan
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Formel:
H₃C(CH₂)₄CH₃ Siedepunkt: 66…69 °C(1,013 hPa) Schmelzpunkt: –95 °C Dichte: 0,660…0.668 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009520 CAS-Nummer: 110-54-3 EINECS: 203-777-6 UN: 1208 ADR: 3,II Merck Index: 13,04712 |
|
Spezifikation Testergebnisse
Assay (GC) | Min. 97.00 % |
Non-volatile matter | Max. 0.0005 % |
Water (Karl Fischer) | Max. 0.010 % |
Free acid (as CH₃COOH) | Max. 0.001 % |
Absorbance at 250 nm | Max. 0.01 |
Absorbance at 225 nm | Max. 0.1 |
Absorbance at 220 nm | Max. 0.7 |
Absorbance at 210 nm | Max. 0.3 |
Absorbance (JP)(Val.Ref) | complying |
IR spectrum | complying |
Peroxides JP | complying |
Vapor Pressure | ~132 mmHg ( 20 °C) |
Refractive Index | n20/D 1.375 (lit.) |
Vapor Density | ~3 (vs air) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...