2-Propanol ≥99,85%, CHROMASOLV™ für die HPLC, Riedel-de Haën™

Lieferant: Honeywell Chemicals
Gefahr

34863-7L 34863-45L 34863-2.5L 34863-1L 34863-4X2.5L 34863-6X1L 34863-185L 34863-18L 34863-30L-RC
34863-7LEA 715.61 EUR
34863-7L 34863-45L 34863-2.5L 34863-1L 34863-4X2.5L 34863-6X1L 34863-185L HONC34863-18L HONC34863-30L-RC
2-Propanol ≥99,85%, CHROMASOLV™ für die HPLC, Riedel-de Haën™
2-Propanol

2-Propanol (Isopropyl alcohol, Isopropanol, IPA) is a colorless solvent and is the simplest example of a secondary alcohol. It can dissolve a wide range of non-polar compounds and works well as a degreaser. Propanol-2 has a boiling point of 82 °C. Suitable for HPLC and spectrophotometry.


  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

Formel: (CH₃)₂CHOH
Siedepunkt: 77…83 °C (1,013 hPa)
Schmelzpunkt: –90 °C
Dichte: 0,784…0.786 g/cm³ (20 °C)
Flammpunkt: 12 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011674
CAS-Nummer: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3,II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

IR spectrum complying
Assay (GC) Min. 99.85 %
Non-volatile matter Max. 0.0003 %
Water (Karl Fischer) Max. 0.05 %
Free acid (as C₂H₅COOH) Max. 0.001 %
Absorbance at 400 nm Max. 0.01
Absorbance at 260 nm Max. 0.01
Absorbance at 230 nm Max. 0.10
Absorbance at 220 nm Max. 0.20
Absorbance at 210 nm Max. 0.70
Absorbance at 205 nm Max. 1.0
APHA Max. 10
Vapor Pressure 33 mmHg ( 20 °C)
Refractive Index n20/D 1.377 (lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR