1-Butanol ≥99,7%, CHROMASOLV™ Plus für die HPLC, Riedel-de Haën™

Lieferant: Honeywell Chemicals
Gefahr

34867-4X2.5L 34867-2.5L 34867-1L 34867-6X1L
34867-4X2.5LEA 697.22 EUR
34867-4X2.5L 34867-2.5L 34867-1L 34867-6X1L
1-Butanol ≥99,7%, CHROMASOLV™ Plus für die HPLC, Riedel-de Haën™
1-Butanol

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities


Beilstein Number: 969148
RTECS Number: 200-751-6
UNSPSC Code: EO1400000

Formel: H₃C(CH₂)₂CH₂OH
Siedepunkt: 119 °C (1,013 hPa)
Schmelzpunkt: <-90 °C
Dichte: 0,81 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002964
CAS-Nummer: 71-36-3
UN: 1120
ADR: 3,III
REACH: 01-2119484630-38
Merck Index: 13,01539
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9395eeb6150a74222da52460f53cedeccd93442&vid=fd8352a556914c5be7213c576355a88e130d1a7b&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Purity (GC) Min. 99.70 %
APHA Max. 10
Non-volatile matter Max. 0.001 %
IR spectrum complying
Water (Karl Fischer) Max. 0.1 %
Free acid (as C₃H₇COOH) Max. 0.002 %
Absorbance at 400 nm Max. 0.01
Absorbance 280 nm Max. 0.01
Absorbance at 260 nm Max. 0.04
Absorbance at 240 nm Max. 0.10
Absorbance at 235 nm Max. 0.10
Absorbance at 225 nm Max. 0.50
Absorbance at 220 nm Max. 0.70
Absorbance at 210 nm Max. 1.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR